Skip to Content
Merck
All Photos(3)

Documents

HS1419A

Pour Boat Weighing Dishes

small, white polystyrene, pack of 500 ea

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.71

material

white polystyrene

packaging

pack of 500 ea

manufacturer/tradename

Heathrow Scientific HS1419A

L × W × H

43 mm × 58 mm × 13 mm

Looking for similar products? Visit Product Comparison Guide

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
The Pour-Boat Weighing Dish is flexible and features smooth surfaces to provide accurate pour-outs with minimal sample loss.

Features and Benefits

  • Design easily dispenses samples without flexing the dish
  • Flat-bottom dishes are made from 12-mil thick polystyrene for better strength, stability, and handling
  • Color contrast to ensure sample is completely removed
  • Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases
  • Temperature range -10 to 70°C (14 to 158°F)
  • Can be used with microwave moisture analyzers
  • FDA/food-grade compliant
  • Disposable
  • Manufactured and packaged in a clean room
  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • pour-spout design easily dispenses samples without flexing the dish
  • flat-bottom dishes are made from 12 mil think polystyrene for better strength, stability, and handling

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service