Key Documents
HS120092
Safety wire bottle carrier
2 x 2.5 lb (bottles), stainless steel
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
stainless steel
manufacturer/tradename
Heathrow Scientific HS120092
capacity
2 x 2.5 lb (bottles)
General description
The Bottle Carrier is perfect for transport of popular sized bottles. You can securely transport two Winchester style bottles up to 135 mm tall and 170 mm diameter (typically 2.5 L capacity).
Choose from one of the most recent versions:
Certificates of Analysis (COA)