Skip to Content
Merck
All Photos(7)

Key Documents

HPA016700

Sigma-Aldrich

Anti-TREML1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-TLT-1, Anti-Trem-like transcript 1 protein precursor, Anti-Triggering receptor expressed on myeloid cells-like protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€543.00

€543.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€543.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€543.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KRKQGNRLGVCGRFLSSRVSGMNPSSVVHHVSDSGPAAELPLDVPHIRLDSPPSFDNTTYTSLPLDSPSGKPSLPAPSSLPPLPPKVLVCSKPVTYATVIFPGGNKGGGTSCGPAQNPPNNQTPSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TREML1(340205)

General description

TREML1 (Triggering receptor expressed on myeloid cells-like 1) is a platelet immunoreceptor membrane protein belonging to the TREM family. It consists of a single Ig superfamily V-type domain and a cytoplasmic tail with two immunoreceptor tyrosine-based inhibition motifs (ITIMs) for inhibitory functions. It is expressed in platelets and alpha-granules of megakaryocytes.

Immunogen

Trem-like transcript 1 protein precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TREML1 (Triggering receptor expressed on myeloid cells-like 1) is majorly involved in the platelet aggregation. It binds to the fibrinogen directly, thereby contributing to the inflammatory responses. It shows inhibitory actions in calcium signaling pathway by recruiting Src homology 2 domain-containing tyrosine phosphatase (SHP)2. Activated soluble TREML1 migrates to the platelet surface during inflammation. It has been suggested that migration of TREML1 may play an important role in vascular hemostasis. Furthermore it accelerates the rate of actin polymerization, platelet aggregation, and adherence to the endothelium during homostasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72294

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander D Barrow et al.
Journal of immunology (Baltimore, Md. : 1950), 172(10), 5838-5842 (2004-05-07)
To date, immunoreceptor tyrosine-based inhibition motifs (ITIMs) have been shown to mediate inhibitory properties. We report a novel triggering receptor expressed on myeloid cells (TREM) family member, TREM-like transcript-1 (TLT1), which differs from the activating members because its cytoplasmic tail
Marc Derive et al.
Journal of immunology (Baltimore, Md. : 1950), 188(11), 5585-5592 (2012-05-04)
The triggering receptor expressed on myeloid cells (TREM)-1 plays a crucial role during the onset of sepsis by amplifying the host immune response. The TREM-like transcript-1 (TLT-1) belongs to the TREM family, is selectively expressed on activated platelets, and is
Jessica Morales et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 21(3), 229-236 (2010-01-23)
Triggering receptor expressed on myeloid cells (TREM) like transcript-1 (TLT-1) is a membrane protein receptor found in alpha-granules of platelets and megakaryocytes. Upon platelet activation TLT-1 is rapidly brought to the surface of platelets. Recently, we demonstrated that activated platelets

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service