Skip to Content
Merck
All Photos(3)

Documents

HPA014859

Sigma-Aldrich

Anti-YIPF3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Killer lineage protein 1, Anti-Natural killer cell-specific antigen KLIP1, Anti-Protein YIPF3, Anti-YIP1 family member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GILDTLEGPNIPPIQRVPRDIPAMLPAARLPTTVLNATAKAVAVTLQSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... YIPF3(25844)

General description

YIPF3 (Yip1 domain family, member 3) is a member of the Yip1 domain family (YIPF), and is a homolog of human Yif1p. It is synthesized in the endoplasmic reticulum (ER) as an N-glycosylated 40kDa protein, which is O-glycosylated to 46kDa protein in the Golgi bodies. Eventually it is cleaved at its luminal C-terminal to produce a 36kDa protein. This protein resides in the cis-Golgi, where it forms a complex with YIPF4. It is a membrane protein, with five transmembrane domains, and is composed of 350 amino acids. This gene is localized to human chromosome 6p21.1-6p21.2, and has nine exons.

Immunogen

Protein YIPF3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

YIPF3 (Yip1 domain family, member 3) forms a complex with YIPF4 in the Golgi apparatus, and is thought to be essential in maintaining the structure of Golgi. Studies suggest that it might be involved in the Golgi to ER (endoplasmic reticulum) retrograde transport. It might also play a role in the trafficking of molecules related to the differentiation and development of vertebrates. It is expressed in nucleated hematopoietic cells, and this is regulated developmentally and ontogenetically. Therefore, this protein might have a role in hematopoiesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73063

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane Prost et al.
Blood cells, molecules & diseases, 29(2), 236-248 (2002-12-20)
A novel membrane protein has been identified in the course of screening for differentially expressed cDNAs in human embryonic hematopoietic sites. This 37- to 38-kDa molecule, designated KLIP-1 (killer lineage protein), consisting of 350 amino acids and containing five transmembrane
Kouji Tanimoto et al.
Cell structure and function, 36(2), 171-185 (2011-07-16)
The Yip1 domain family (YIPF) proteins are homologues of yeast Yip1p and Yif1p, which are proposed to function in ER to Golgi transport. Here, we report the characterization of YIPF3 and YIPF4, homologues of human Yif1p and Yip1p, respectively. Immunofluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service