Skip to Content
Merck
All Photos(2)

Documents

H3166

Sigma-Aldrich

Anti-HOXD4 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-HHO.C13, Anti-HOX-4B, Anti-HOX-5.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~35 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 1.5-3 μg using lysates of HEK-293T cells over-expressing human HOXD4
indirect immunofluorescence: 0.25-0.5 μg/mL using paraformaldehyde fixed HEK-293T cells over expressing human HOXD4
western blot: 0.5-1 μg/mL using lysates of HEK-293T cells over-expressing human HOXD4

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXD4(3233)
rat ... Hoxd4(288153)

General description

The Homeobox D4 (HOXD4) gene is expressed in mesodermal tissues and the central nervous system (CNS). The HOXD4 gene is mapped on the human chromosome at 2q31.1.

Specificity

Anti-HOXD4 (C-terminal) recognizes human HOXD4 (also known as HOX-4B, HOX-5).

Application

Anti-HOXD4 (C-terminal) antibody produced in rabbit may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

The Homeobox D4 (HOXD4) gene is involved in the development of dura mater and morphogenesis of the development of the spine. This gene plays a role in the pathogenesis of spinal extradural arachnoid cyst (SEDAC). HOXD4 gene is also involved in tumorigenesis of the central nervous system. This gene acts as a diagnostic marker of gliomas.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin-Wei Zhao et al.
Oncotarget, 8(70), 115657-115666 (2018-02-01)
HOX (homologous box) is known as the dominant gene of vertebrate growth and cell differentiation. Abnormal expression of HOX gene in various tumors has attracted the attention of scholars. As a component of HOX clusters, HOXD4 plays a controversial role
Hui Liu et al.
Saudi journal of gastroenterology : official journal of the Saudi Gastroenterology Association, 25(1), 46-54 (2018-12-28)
Homeobox D4 (HOXD4) belongs to the homeobox (HOX) family, which plays a crucial role in the early embryo development and cell differentiation. The role of HOXD4 in human gastric adenocarcinoma has not been elucidated. In the present study, we aimed
Yoji Ogura et al.
PloS one, 10(11), e0142126-e0142126 (2015-11-07)
Spinal extradural arachnoid cyst (SEDAC) is a cyst in the spinal canal that protrudes into the epidural space from a defect in the dura mater and leads to neurological disturbances. We previously showed that familial SEDAC is caused by FOXC2
Mojgan Rastegar et al.
Molecular and cellular biology, 24(18), 8090-8103 (2004-09-02)
Hox genes are differentially expressed along the embryonic anteroposterior axis. We used chromatin immunoprecipitation to detect chromatin changes at the Hoxd4 locus during neurogenesis in P19 cells and embryonic day 8.0 (E8.0) and E10.5 mouse embryos. During Hoxd4 induction in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service