Skip to Content
Merck
All Photos(1)

Key Documents

GW10807F

Sigma-Aldrich

Anti-GAG polyprotein antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-GAG_HV1N5, Anti-Gag polyprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

viral

manufacturer/tradename

Genway 15-288-10807F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human immunodeficiency virus 1 ... Gag(155030)

Immunogen

Immunogen Sequence: GI # 12831136, sequence 2-132
Recombinant gag polyprotein (HIV-1 vector pNL4-3)

Application

Anti-GAG polyprotein antibody produced in chicken is suitable for western blot analysis.

Biochem/physiol Actions

GAG is highly involved in RNA trafficking. It helps in transport of human immunodeficiency virus type 1 (HIV-1) RNA via diffusion within the cytoplasm. Ubiquitination of Gag enhances possibilities of its optimal function in virus release and maturation. It also recruits Ub ligases at the viral assembly site.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianbo Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(48), E5205-E5213 (2014-11-19)
Full-length HIV-1 RNA plays a central role in viral replication by serving as the mRNA for essential viral proteins and as the genome packaged into infectious virions. Proper RNA trafficking is required for the functions of RNA and its encoded
U Schubert et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(24), 13057-13062 (2000-11-23)
Retrovirus assembly and maturation involve folding and transport of viral proteins to the virus assembly site followed by subsequent proteolytic cleavage of the Gag polyprotein within the nascent virion. We report that inhibiting proteasomes severely decreases the budding, maturation, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service