Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5167-01

Sigma-Aldrich

Mono Q® 10/100 GL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

ligand

-OCH2CH(OH)CH2OCH2CH(OH)CH2N+(CH3)3

material

PEEK column

description

Ion Exchanger Type (value)

packaging

pkg of 1 ea

parameter

580 psi

bed volume

8 mL

column I.D.

10 mm

matrix

monodisperse porous polystyrene/divinyl benzene beads
polymeric

avg. part. size

10 μm

working range

2-12

separation technique

anion exchange

General description

Mono Q® is a strong anion exchanger prepacked with MonoBeads in a Tricorncolumn. The column is an excellent choice for small-scale polishing in purification of proteins, peptides, and other biomolecules when high purity is required.

The Tricorn columns offer high-resolution separations with greater loading capacity but slightly lower resolution than MiniBeads columns.

Features and Benefits

  • Monodisperse 10 μm porous beads yield high resolution, dynamic capacity, reproducibility, and durability.
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers.
  • Compatible with AKTA design, FPLC System, and other high-performance LC systems.
  • MonoBeads are also available as Mono P® for chromatofocusing

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

FPLC is a trademark of Cytiva
Mono P is a registered trademark of Cytiva
Mono Q is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.

Protocols

This page covers using Monobeads for purification of proteins, peptides, or oligonucleotides.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service