Key Documents
GE11-0007-68
Solvent Filter
(PTFE), Cytiva
About This Item
material
(PTFE)
description
The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100% Ethyl acetate, 100% Hexane and 100% Tetrahydrofuran
feature
Wetted Material: Polytetrafluoroethene (PTFE)
manufacturer/tradename
11-0007-68
Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)