Key Documents
GBL705003
Grace Bio-Labs ONCYTE® SuperNOVA nitrocellulose film slides
3-18 mm x 16 mm NC pads per slide, plain glass
Synonym(s):
nitrocellulose slides, protein microarrays
Select a Size
20 EA
€790.00
€790.00
Please contact Customer Service for Availability
Select a Size
Change View
20 EA
€790.00
About This Item
UNSPSC Code:
41122600
NACRES:
NB.15
€790.00
Please contact Customer Service for Availability
material
nitrocellulose (on glass)
packaging
pkg of 20 ea
manufacturer/tradename
Grace Bio-Labs 705003
external size
25 mm × 75 mm
binding type
high binding surface
General description
Fluorescence detection is a preferred method for many quantitative protein microarray assaysin research and diagnostic applications due to its excellent sensitivity and potential for multiplexing. Use of multiplexed fluorescence-based assays on porous nitrocellulose though has been hampered by high backgrounds at shorter excitation wave lengths (such as the 532nm fluorescence channel). These ONCYTE™ SuperNOVA™ microporous nitrocellulose film slides increase protein microarray assay sensitivity and dynamic range using fluorescent end points at multiple wave lengths. Use of SuperNOVA™ slides can lower the limit of detection with fluorescent end points by 3 – 4 orders of magnitude over competing nitrocellulose film slides.
Legal Information
NOVA is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
SuperNOVA is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)