Skip to Content
Merck
All Photos(1)

Documents

G9502

Sigma-Aldrich

Guanylyl(3′→5′)uridine ammonium salt

Synonym(s):

GpU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H24N7O13P · NH3
CAS Number:
Molecular Weight:
606.44
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

N.NC1=NC(=O)c2ncn(C3OC(CO)C(OP(O)(=O)OCC4OC(C(O)C4O)N5C=CC(=O)NC5=O)C3O)c2N1

InChI

1S/C19H24N7O13P.H3N/c20-18-23-14-9(15(32)24-18)21-5-26(14)17-12(31)13(6(3-27)37-17)39-40(34,35)36-4-7-10(29)11(30)16(38-7)25-2-1-8(28)22-19(25)33;/h1-2,5-7,10-13,16-17,27,29-31H,3-4H2,(H,34,35)(H,22,28,33)(H3,20,23,24,32);1H3

InChI key

BBBUJXLTQPKTLN-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hande Boyaci et al.
Nature, 565(7739), 382-385 (2019-01-11)
A key regulated step of transcription is promoter melting by RNA polymerase (RNAP) to form the open promoter complex1-3. To generate the open complex, the conserved catalytic core of the RNAP combines with initiation factors to locate promoter DNA, unwind
James Chen et al.
Molecular cell, 68(2), 388-397 (2017-10-11)
Noncoding RNAs (ncRNAs) regulate gene expression in all organisms. Bacterial 6S RNAs globally regulate transcription by binding RNA polymerase (RNAP) holoenzyme and competing with promoter DNA. Escherichia coli (Eco) 6S RNA interacts specifically with the housekeeping σ
Alena Kroupova et al.
Nucleic acids research, 47(2), 1030-1042 (2018-11-22)
Non-templated 3'-uridylation of RNAs has emerged as an important mechanism for regulating the processing, stability and biological function of eukaryotic transcripts. In Drosophila, oligouridine tailing by the terminal uridylyl transferase (TUTase) Tailor of numerous RNAs induces their degradation by the
Elizabeth Davis et al.
Nucleic acids research, 43(1), 433-445 (2014-12-17)
Escherichia coli has served as the archetypal organism on which the overwhelming majority of biochemical characterizations of bacterial RNA polymerase (RNAP) have been focused; the properties of E. coli RNAP have been accepted as generally representative for all bacterial RNAPs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service