Skip to Content
Merck
All Photos(3)

Key Documents

G7018

Sigma-Aldrich

α-D-Glucose 1-phosphate disodium salt hydrate

≥98%, BioXtra, lyophilized powder

Synonym(s):

α-D-Glucopyranose 1-phosphate, Cori ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11O9PNa2 · xH2O
CAS Number:
Molecular Weight:
304.10 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
To inquire about this product G7018, please contact your local Merck office or dealer. Contact Technical Service

biological source

synthetic

product line

BioXtra

Assay

≥98%

form

lyophilized powder

impurities

glucose, starch, and ethanol, essentially free
0.01-0.2% α-D-glucose 1,6-diphosphate

color

white

solubility

water: 50 mg/mL, clear

cation traces

Na: 13.7-17.1% (anhydrous)

storage temp.

−20°C

SMILES string

[Na+].[Na+].[H]O[H].OC[C@H]1O[C@H](OP([O-])([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H13O9P.2Na.H2O/c7-1-2-3(8)4(9)5(10)6(14-2)15-16(11,12)13;;;/h2-10H,1H2,(H2,11,12,13);;;1H2/q;2*+1;/p-2/t2-,3-,4+,5-,6-;;;/m1.../s1

InChI key

YWAUQXHOCBBYLP-FBNUBEQJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

α-D-Glucose 1-phosphate is converted into D-glucose-6-phosphate by the enzyme phosphoglucomutase.

Biochem/physiol Actions

α-D-Glucose 1-phosphate is the α-anomeric form of glucose which contains a phosphate group on the primary carbon. In certain strains of Corynebacteria, glucose 1-phosphate has been shown to induce the build up of starch-like inclusion bodies within the cell. [1]

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose-1-Phosphate-Induced Accumulation of Intracellular Starch: a Distinguishing Feature of Certain Corynebacteria.
Arden, S.
International J. Systemic Bact., 24, 139-141 (1974)
Rickard Olsson et al.
Environmental science & technology, 46(1), 285-291 (2011-11-23)
Esters of phosphoric acid constitute a sizable fraction of the total phosphorus supply in the environment and thus play an important role in the global phosphorus cycle. Enzymatic hydrolysis of these esters to produce orthophosphate is often a required reaction
Xingwei Chen et al.
Journal of biomaterials applications, 27(4), 391-402 (2011-07-14)
The purpose of this study was to evaluate the feasibility of in situ thermosensitive hydrogel based on chitosan in combination with disodium α-d-Glucose 1-phosphate (DGP) for ocular drug delivery system. Aqueous solution of chitosan/DGP underwent sol-gel transition as temperature increased
Lyann Sim et al.
The FEBS journal, 280(4), 1073-1083 (2013-01-04)
The mechanism of yeast flocculation is generally considered to be mediated through the interaction of cell surface flocculins and mannan carbohydrates. In the present study, the crystal structure of the soluble 25-kDa lectin domain of flocculin 1 from brewer's yeast
Rickard Olsson et al.
Langmuir : the ACS journal of surfaces and colloids, 26(24), 18760-18770 (2010-11-20)
Adsorption, desorption, and precipitation reactions at environmental interfaces govern the fate of phosphorus in terrestrial and aquatic environments. Typically, a substantial part of the total pool of phosphorus consists of organophosphate, and in this study we have focused on the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service