Skip to Content
Merck
All Photos(1)

Key Documents

G6535

Sigma-Aldrich

Arg-Arg-Gastrin Fragment 22-30 human

Synonym(s):

Arg-Arg-Leu-Glu-Glu-Glu-Glu-Glu-Ala-Tyr-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H89N17O23
CAS Number:
Molecular Weight:
1380.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](N)CCCNC(N)=N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)NCC(O)=O

InChI

1S/C57H89N17O23/c1-27(2)24-38(74-50(92)32(7-5-23-64-57(61)62)67-47(89)31(58)6-4-22-63-56(59)60)55(97)72-37(16-21-44(84)85)54(96)71-36(15-20-43(82)83)53(95)70-35(14-19-42(80)81)52(94)69-34(13-18-41(78)79)51(93)68-33(12-17-40(76)77)49(91)66-28(3)46(88)73-39(48(90)65-26-45(86)87)25-29-8-10-30(75)11-9-29/h8-11,27-28,31-39,75H,4-7,12-26,58H2,1-3H3,(H,65,90)(H,66,91)(H,67,89)(H,68,93)(H,69,94)(H,70,95)(H,71,96)(H,72,97)(H,73,88)(H,74,92)(H,76,77)(H,78,79)(H,80,81)(H,82,83)(H,84,85)(H,86,87)(H4,59,60,63)(H4,61,62,64)/t28-,31-,32-,33-,34-,35-,36-,37-,38-,39-/m0/s1

InChI key

IZPALEYOHFCFHY-UYZDLLKLSA-N

Substrates

Substrate for tyrosine kinase

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphorylation of a synthetic gastrin peptide by the tyrosine kinase of A431 cell membranes.
G S Baldwin et al.
Biochemical and biophysical research communications, 109(3), 656-663 (1982-12-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service