Skip to Content
Merck
All Photos(1)

Key Documents

G2138

Sigma-Aldrich

rac-Glycerol 1-phosphate disodium salt hydrate

≥85% (enzymatic)

Synonym(s):

DL-α-Glycerophosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7Na2O6P · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
Beilstein:
5177716
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥85% (enzymatic)

storage temp.

2-8°C

SMILES string

O.[Na+].[Na+].OCC(O)COP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(5)2-9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

OFNNKPAERNWEDD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

rac-Glycerol 1-phosphate does not affect the growth and metabolism of E. coli at concentrations of 5-100μm.

Biochem/physiol Actions

rac-Glycerol 1-phosphate was shown to be produced in mammalian tissues by the hydrolysis of rac-glycerol 1:2-cyclic phosphate by a soluble phosphodiesterase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiangwei Yao et al.
Biochimica et biophysica acta, 1831(3), 495-502 (2012-09-18)
Membrane phospholipid synthesis is a vital facet of bacterial physiology. Although the spectrum of phospholipid headgroup structures produced by bacteria is large, the key precursor to all of these molecules is phosphatidic acid (PtdOH). Glycerol-3-phosphate derived from the glycolysis via
Kyung Ok Yu et al.
Biotechnology and bioengineering, 110(1), 343-347 (2012-08-14)
Triacylglycerol (TAG) is a microbial oil feedstock for biodiesel production that uses an inexpensive substrate, such as glycerol. Here, we demonstrated the overproduction of TAG from glycerol in engineered Saccharomyces cerevisiae via the glycerol-3-phosphate (G3P) pathway by overexpressing the major
Luana Lopes Souza et al.
The Journal of endocrinology, 211(1), 65-72 (2011-07-15)
n-3 polyunsaturated fatty acids (n-3 PUFA) from fish oil (FO) exert important lipid-lowering effects, an effect also ascribed to thyroid hormones (TH) and TH receptor β1 (TRβ1)-specific agonists. n-3 PUFA effects are mediated by nuclear receptors, such as peroxisome proliferator-activated
Heinrich Topp et al.
Pharmacology, 88(3-4), 193-200 (2011-10-12)
The primary aim of the present investigation was to determine and compare the pharmacokinetic (PK) profiles of inorganic phosphate in serum and urine after intravenous administration of sodium glycerophosphate and inorganic sodium phosphate. Additionally, study product safety profiles were evaluated.
Adam L Orr et al.
The Journal of biological chemistry, 287(51), 42921-42935 (2012-11-06)
The oxidation of sn-glycerol 3-phosphate by mitochondrial sn-glycerol 3-phosphate dehydrogenase (mGPDH) is a major pathway for transfer of cytosolic reducing equivalents to the mitochondrial electron transport chain. It is known to generate H(2)O(2) at a range of rates and from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service