G1663
Glycogen Synthase Kinase 3β, Histidine-tagged from rabbit
≥90% (SDS-PAGE), recombinant, expressed in E. coli
Synonym(s):
GSK-3β, Kinase FA, Tau protein kinase I
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
recombinant
expressed in E. coli
Assay
≥90% (SDS-PAGE)
form
lyophilized solid
specific activity
5000.00-30000.00 units/mg protein
mol wt
46,712 Da
relevant disease(s)
Alzheimer′s disease
shipped in
dry ice
storage temp.
−20°C
General description
Glycogen Synthase Kinase 3β (GSK3β) is one of the two isoforms of GSK3 enzyme, both of which are encoded by different genes. This protein is predominantly cytoplasmic, but is also found to be localized to nucleus and mitochondria. It is a protein-serine kinase. This protein contains a protein kinase domain in its core, and is composed of 482 amino acids with a molecular weight of 46,712Da. It shows a wide range of tissue expression with the highest expression in brain.
Application
Glycogen Synthase Kinase 3β (GSK3β) has been used
- to study GSK3β cleavage by calpain
- to perform motility assay on squid axoplasms
- for the pre-incubation of progesterone receptor A during in vitro ubiquitination assay
Biochem/physiol Actions
Glycogen Synthase Kinase 3β (GSK3β) is responsible for rapid cyclin D1 turnover by phosphorylating it on Thr-286. It functions as a negative regulator of Wnt signaling during embryonic development cell proliferation in adult tissues. This protein phosphorylates and regulates multiple proteins, such as glycogen synthase, insulin receptor substrate-1, axin and APC (adenomatous polyposis coli). This protein is also shown to interact with multiple proteins, which are linked with Alzheimer′s disease (AD).
Unit Definition
One unit will transfer one pmole of phosphate from ATP to phosphatase inhibitor 2 per min at pH 7.5 at 30 °C.
Reconstitution
Reconstitution to 100 μl with deionized water produces a solution in 20 mM Tris, pH 7.5, 2 mM EDTA, 2 mM EGTA, 1 mM DTT, 5% sucrose, 0.1% Brij® L23, 1 μg/ml leupeptin, and 0.1 mM AEBSF.
Legal Information
Brij is a registered trademark of Croda International PLC
inhibitor
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Enzymes, 17, 461-461 (1986)
The Biochemical journal, 288 ( Pt 1), 309-314 (1992-11-15)
Multifunctional ATP-citrate lyase kinase (ACLK) exhibits several properties that are similar to glycogen-synthase kinase-3 (GSK-3). The molecular cloning of two distinct mammalian GSK-3 cDNAs and a Drosophila melanogaster (fruitfly) homologue, zeste-white3sgg, has established the existence of a GSK-3 subfamily. A
Journal of protein chemistry, 14(2), 95-105 (1995-02-01)
Previously, tau protein kinase I/glycogen synthase kinase-3 beta/kinase FA(TPKI/GSK-3 beta/FA) was identified as a brain microtubule-associated tau kinase possibly involved in the Alzheimer disease-like phosphorylation of tau. In this report, we find that the TPKI/GSK-3 beta/FA can be stimulated to
The Journal of biological chemistry, 288(36), 26265-26274 (2013-07-25)
Germ line mutations of the BRCA1 gene increase the risk of breast and ovarian cancer, but the basis of this tissue-specific tumor predisposition is not fully understood. Previously, we reported that the progesterone receptors are stabilized in Brca1-deficient mammary epithelial
The Journal of biological chemistry, 269(20), 14566-14574 (1994-05-20)
The enzyme glycogen synthase kinase-3 (GSK-3) has been implicated in the control of several metabolic enzymes and transcription factors in response to extracellular signals. In the past, the enzyme has been considered to be a protein Ser/Thr kinase although it
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service