Key Documents
F5051
Corning® FiltrEX 96 well filter plates
96 well, PVDF membrane, pore size 0.2 μm
About This Item
UNSPSC Code:
41104900
material
PVDF membrane
polystyrene
rectangular bottom white polystyrene wells
white polystyrene plate
sterility
sterile
feature
lid: no
plate format: 96 well filter plate, standard
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3505
maximum volume
350 μL
working volume
350 μL
pore size
0.2 μm
General description
Glass fiber filter plates (CLS3510, CLS3511) can be used for a variety of applications, including plasmid isolation, DNA purification, or receptor/ligand binding assays. The low-binding hydrophilic PVDF membrane filter plates (CLS3504, CLS3505) are suitable for protein kinase assays or for bead and resin based separation assays.
Legal Information
Corning is a registered trademark of Corning, Inc.
FiltrEX is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)