EP5409708007
Rotor for Eppendorf® 5427R Centrifuge
FA-45-30-11, non aerosol-tight, to hold, 30 x 1.5/2.0 mL tubes, with rotor lid
All Photos(1)
About This Item
Recommended Products
feature
to hold 30 x 1.5/2.0 mL tubes
fixed angle rotor
non aerosol-tight
with rotor lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5409708007 (Global)
parameter
14,000 rpm
20,800 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5427R
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG