Key Documents
EP5242956003
Eppendorf® Microloader
pkg of 192 ea
Synonym(s):
Flow indicator
About This Item
UNSPSC Code:
41121600
packaging
pkg of 192 ea
manufacturer/tradename
Eppendorf® 5242956003
General description
  • Microloader tips are extremely long, fine, and flexible for filling microcapillaries for microinjection
  • Unique tips are ideal for recovering surplus solution from the capillary
  • Provides the ideal solution where additional reach is needed while pipetting the smallest volumes.
Features and Benefits
  • Microloader tip with a volume range of 0.5-20 uL
  • Light gray color tips of 100mm length
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Microloader is a trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)