Key Documents
EP3116000112
Accessories for Eppendorf® Pipettes & Repeaters
Pipette Holder 2, for one Eppendorf Research™, Eppendorf Research™ plus, Reference™, Reference™ 2 or Biomaster™
Select a Size
1 EA
€35.60
€35.60
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€35.60
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
€35.60
Please contact Customer Service for Availability
feature
for one Eppendorf Research, Eppendorf Research plus, Reference, Reference 2 or Biomaster
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 3116000112 (Global)
Related Categories
General description
Accessories for Eppendorf pipettes and repeaters,
Eppendorf® provides holders suitable for all current manual and electronic liquid handling instruments as well as most predecessor manual pipettes. These holders allow you to have your pipette and Multipette at convenient locations, such as mounted to a wall, on a shelf above the bench, or inside a biological safety cabinet.
Features and Benefits
  • Compatible for one Eppendorf Research®, Eppendorf Research® plus, Eppendorf Reference®, Eppendorf Reference® 2 or Biomaster®
  • for Pipette Carousel 2 and Charger Carousel 2 or wall mounting, sticky tape is included for effortless installation
Legal Information
Biomaster is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Research is a registered trademark of Eppendorf AG
Reference is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)