Skip to Content
Merck
All Photos(1)

Key Documents

EMU043481

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Serpinh1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGCGAACACTCCAAGATCAACTTCCGAGACAAGCGCAGCGCCCTGCAGTCCATCAACGAGTGGGCCTCGCAGACCACGGACGGCAAGCTGCCTGAGGTCACCAAGGATGTGGAGCGCACGGATGGGGCACTGCTTGTGAACGCCATGTTCTTTAAGCCACACTGGGATGAGAGGTTTCACCACAGGATGGTGGACAACCGTGGCTTCATGGTGACCCGCTCCTATACTGTGGGTGTTACGATGATGCACCGGACAGGCCTGTACAACTACTATGACGACGAGAAGGAGAAGCTGCAGATGGTGGAGATGCCCCTGGCTCACAAGCTCTCCAGCCTCATCATCCTCATGCCCCACCATGTGGAGCCGCTCGAGCGCTTGGAGAAGCTGCTGACCAAGGAGCAGCTGAAGGCCTGGATGGGAAAGATGCAGAAGAAGGCTGTCGCCATCTCCCTGCCCAAGGGCGTGGTGGAGGTGACCCATGACCTGCAGAAACATCTGGCAGGACTGGGCCTGACCGAAGCCATCGACAAGAACAAGGCAGACCTATCGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingga Morry et al.
Biomaterials, 66, 41-52 (2015-07-22)
Fibrotic diseases such as scleroderma have been linked to increased oxidative stress and upregulation of pro-fibrotic genes. Recent work suggests a role of NADPH oxidase 4 (NOX4) and heat shock protein 47 (HSP47) in inducing excessive collagen synthesis, leading to
Haiyan Chu et al.
Protein & cell, 6(8), 589-598 (2015-06-21)
Uncontrolled fibrosis of skin and internal organs is the main characteristic of scleroderma, and collagen is a major extracellular matrix protein that deposits in the fibrotic organs. As the chaperone of collagen, heat shock protein 47 (HSP47) is closely related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service