Skip to Content
Merck
All Photos(1)

Key Documents

EHU144921

Sigma-Aldrich

MISSION® esiRNA

targeting human HMGCS2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCGAAAGTGTGTGTCTCCTGAGGAGTTCACAGAAATAATGAACCAAAGAGAGCAATTCTACCATAAGGTGAATTTCTCCCCACCTGGTGACACAAACAGCCTTTTCCCAGGTACTTGGTACCTGGAGCGAGTGGACGAGCAGCATCGCCGAAAGTATGCCCGGCGTCCCGTCTAAAGGTGTTCTGCAGATCCATGGAAAGCTTCCTGGGAAACGTATGCTAGCAGAGCTTCTCCCCGTGAATCATATTTTTAAGATCCCACTCTTAGCTGGTAAATGAATTTGAATCGACATAGTAGCCCCATAAGCATCAGCCCTGTAGAGTGAGGAGCCATCTCTAGCGGGCCCTTCATTCCTCTCCATGCTGCAATCACTGTCCTGGGCTTATGGTGCTATGGACTAGGGGTCCTTTGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Le Shi et al.
Neurobiology of aging, 56, 115-126 (2017-05-23)
Alzheimer's disease (AD) is considered a multifactorial disease that affects the central nervous system and periphery. A decline in brain glucose metabolism is an early feature of AD and is accompanied by a phenotypic shift from aerobic glycolysis to ketogenesis.
Li Wang et al.
IUBMB life, 72(9), 1997-2009 (2020-08-01)
Diabetic cardiomyopathy (DCM) is a cardiac disorder, which affects around 12% diabetic patients, resulting in overt heart death. Our initial bioinformatic analysis identified the differentially expressed gene 3-hydroxy-3-methylglutaryl-coenzyme A synthase 2 (HMGCS2) in DCM, which may be activated by peroxisome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service