Skip to Content
Merck
All Photos(1)

Documents

EHU116371

Sigma-Aldrich

MISSION® esiRNA

targeting human ANXA2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CATGTTGGAAAGCATCAGGAAAGAGGTTAAAGGAGACCTGGAAAATGCTTTCCTGAACCTGGTTCAGTGCATTCAGAACAAGCCCCTGTATTTTGCTGATCGGCTGTATGACTCCATGAAGGGCAAGGGGACGCGAGATAAGGTCCTGATCAGAATCATGGTCTCCCGCAGTGAAGTGGACATGTTGAAAATTAGGTCTGAATTCAAGAGAAAGTACGGCAAGTCCCTGTACTATTATATCCAGCAAGACACTAAGGGCGACTACCAGAAAGCGCTGCTGTACCTGTGTGGTGGAGATGACTGAAGCCCGACACGGCCTGAGCGTCCAGAAATGGTGCTCACCATGCTTCCAGCTAACAGGTCTAGAAAACCAGCTTGCGAATAACAGTCCCCGTGGCCATCCCTGTGAGGGTGACGTTAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanhong Chang et al.
Oncology reports, 40(6), 3625-3634 (2018-10-03)
Recently, long non-coding RNA (lncRNA) FOXD2 adjacent opposite strand RNA 1 (FOXD2-AS1) has been recognized to function as an oncogene in several human tumors, and FOXD2‑AS1 dysregulation has been closely associated with carcinogenesis and tumor progression. Nevertheless, the correlation between the
Yan Liu et al.
Oncology reports, 37(6), 3643-3650 (2017-04-26)
Annexin A2 is a member of the Annexin family that acts as a Ca2+-dependent phospholipid and membrane binding protein, which is associated with the survival and spread of multiple neoplasms. However, the function of Annexin A2 in ovarian cancer progression remains unclear.
Weining Wu et al.
Journal of experimental & clinical cancer research : CR, 38(1), 133-133 (2019-03-23)
Glioblastoma multiforme (GBM) is the most common and aggressive form of astrocytoma among adult brain tumors. Multiple studies have shown that long non-coding RNAs (lncRNAs) play important roles in acting as molecular sponge for competing with microRNAs (miRNAs) to regulate
Jie Liu et al.
BMC biochemistry, 4, 10-10 (2003-09-10)
Annexin II heavy chain (also called p36, calpactin I) is lost in prostate cancers and in a majority of prostate intraepithelial neoplasia (PIN). Loss of annexin II heavy chain appears to be specific for prostate cancer since overexpression of annexin
Isah Abubakar Aliyu et al.
Viruses, 11(4) (2019-04-12)
Recent evidence has demonstrated that dengue virus requires active filopodia formation for a successful infection. However, the cellular factor involved in the interaction has not been fully elucidated. We used a combination of virus overlay protein binding assay and LC-MS/MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service