Skip to Content
Merck
All Photos(1)

Key Documents

EHU059281

Sigma-Aldrich

MISSION® esiRNA

targeting human TCF12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATCCTGGGCTTAGTGAAACTACCAACCCTATGGGTCATATGTAAACATCAGCCAGTTCCAGAGTTATCAGTAGGCTAGATAGAAGGTGACCTCTCCTCATAAGGACTTGGACAACTCAGATTATCTGAAGACACAAACCTGACAGGAGGGAGAAGAAAAAACAAAACACTTGAACCAAGAAACTCAAATGTAATCCTACGATCAAAGCAACTGGTCAACACTTCCATCAGAAGTGAAGATAGGAAGCTCATCAGATAGAACATCAGCCCATGAGATGTTTGCAACAAATCTTTTGTTGCAAGCAGTGTGTCGCTTCTGCACAATCAGAGACTGTCTCGATCTCTCCACTCACCGTGGAAGTTGCCTTGTGCCTAAACTGAATTGACAAATGCATTGTAACTACAAATTTTATTTATTGTTATGAAACTGTAAGGTCTACATATAAAGGGAAAAAGTTAATGTGGAAAGCTGATCTACACTCAGCTGATGCCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paulo R D V Godoy et al.
Molecular medicine reports, 14(6), 5253-5260 (2016-10-26)
Glioblastoma multiforme (GBM) is a lethal tumor and novel strategies are required to overcome resistance. Transcription factor 12 (HEB) has been associated with neural and stem cell proliferation, is overexpressed in certain tumor types and is induced in irradiated U87MG cells.
Leila Pirhaji et al.
Nature communications, 8(1), 623-623 (2017-09-22)
The immense and growing repositories of transcriptional data may contain critical insights for developing new therapies. Current approaches to mining these data largely rely on binary classifications of disease vs. control, and are not able to incorporate measures of disease
Duo Wen et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(8), 6211-6221 (2015-03-11)
Malic enzyme 1 (ME1) links the glycolytic and citric acid cycles and is important for NADPH production, glutamine metabolism, and lipogenesis. Recently, its deregulation has been implicated in the progression of various cancers. However, the role of ME1 in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service