Skip to Content
Merck
All Photos(1)

Key Documents

E6136

Sigma-Aldrich

α-Endorphin human

≥97% (HPLC)

Synonym(s):

β-Lipotropin (61-76), human, synthetic, β-Lipotropin 61-76, LPH (61-76), Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C77H120N18O26S
CAS Number:
Molecular Weight:
1745.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CSCC[C@H](NC(=O)[C@H](Cc1ccccc1)NC(=O)CNC(=O)CNC(=O)[C@@H](N)Cc2ccc(O)cc2)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CO)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N3CCC[C@H]3C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H]([C@@H](C)O)C(O)=O

InChI

1S/C77H120N18O26S/c1-38(2)31-51(70(113)91-60(39(3)4)74(117)94-63(42(7)100)77(120)121)88-73(116)55-18-14-29-95(55)76(119)62(41(6)99)93-67(110)48(23-25-56(80)102)85-71(114)53(36-96)89-65(108)47(17-12-13-28-78)84-66(109)49(24-26-59(105)106)86-72(115)54(37-97)90-75(118)61(40(5)98)92-68(111)50(27-30-122-8)87-69(112)52(33-43-15-10-9-11-16-43)83-58(104)35-81-57(103)34-82-64(107)46(79)32-44-19-21-45(101)22-20-44/h9-11,15-16,19-22,38-42,46-55,60-63,96-101H,12-14,17-18,23-37,78-79H2,1-8H3,(H2,80,102)(H,81,103)(H,82,107)(H,83,104)(H,84,109)(H,85,114)(H,86,115)(H,87,112)(H,88,116)(H,89,108)(H,90,118)(H,91,113)(H,92,111)(H,93,110)(H,94,117)(H,105,106)(H,120,121)/t40-,41-,42-,46+,47+,48+,49+,50+,51+,52+,53+,54+,55+,60+,61+,62+,63+/m1/s1

InChI key

NXSIJWJXMWBCBX-NWKQFZAZSA-N

Gene Information

human ... POMC(5443)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-Ser-Gln-Thr-Pro-Leu-Val-Thr

General description

α-Endorphin is one of the three endorphins found in the brain, which has morphine-mimetic activities. It is a hexadecapeptide with a primary structure of H-Tyr-Gly-Gly-Phe-Met-Thr-Ser-Glu-Lys-SerGln-Thr-Pro-Leu-Val-Thr-OH.[1] It is a fragment of β-Endorphin (β-LPH61-91), which is an endogenous opioid ligand, and is denoted as (β-LPH61-76).[2] It is found in the hypothalamus and pituitary.[3]

Biochem/physiol Actions

α-Endorphin is a substrate for transglutaminase[4], and has stimulatory effects on immune system function.[5] It influences the self-stimulating tendencies in rat, related to electrical stimulation of the ventral tegmentum area neurons. It counteracts the effect of γ-endorphin and promotes this behavior in rats.[6] The functionality of α-endorphin is comparable to that of psychostimulant drugs, and as it acts in an opposing manner to γ-endorphin, these peptides might have a central role in the homeostasis of brain function.[2]

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

gamma endorphin, alpha endorphin and Met-enkephalin are formed extracellularly from lipotropin C fragment.
B M Austen et al.
Nature, 269(5629), 619-621 (1977-10-13)
J P Huidobro-Toro et al.
The Journal of pharmacology and experimental therapeutics, 222(1), 262-269 (1982-07-01)
The inhibitory activity of a variety of opioid peptides was tested on the electrically stimulated rat vas deferens. Human beta-endorphin (beta h-EP) was the most potent of the opoioid-like peptides; it produced half-maximal inhibition of the neuromuscular twitching at a
J P Burbach et al.
The Journal of biological chemistry, 260(11), 6663-6669 (1985-06-10)
The peptides that represent the major components with alpha-endorphin- and gamma-endorphin-like immunoreactivity in the rat neurointermediate lobe were purified to homogeneity and chemically characterized. Rat neurointermediate lobes were extracted by boiling and homogenization in acetic acid. Peptide purification was based
N Ling et al.
Proceedings of the National Academy of Sciences of the United States of America, 73(11), 3942-3946 (1976-11-01)
The isolation and primary structure of two peptides with morphinomimetic activity, obtained from an extract of porcine hypothalamus-neurohypophysis, are described. The amino acid sequence of the two peptides, named alpha-endorphin and gamma-endophin, was determined by mass spectrometry and danxyl-Edman methods
A A Waterfield et al.
European journal of pharmacology, 58(1), 11-18 (1979-09-01)
For characterisation in vitro, four parallel assays were used: the guinea-pig ileum and mouse vas deferens as pharmacological models at 36 degrees C and the inhibition of binding of [3H]-naltrexone, [3H]-leucine-enkephalin and [3H]-methione-enkephalin at 0 degrees C. The Leu65-analogue of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service