Skip to Content
Merck
All Photos(1)

Key Documents

E3518

Sigma-Aldrich

Ethylenediaminetetraacetic acid triethyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28N2O8 · xHCl
CAS Number:
Molecular Weight:
376.40 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

SMILES string

Cl.CCOC(=O)CN(CCN(CC(=O)OCC)CC(=O)OCC)CC(O)=O

InChI

1S/C16H28N2O8.ClH/c1-4-24-14(21)10-17(9-13(19)20)7-8-18(11-15(22)25-5-2)12-16(23)26-6-3;/h4-12H2,1-3H3,(H,19,20);1H

InChI key

YWHJXEBZSBYHLB-UHFFFAOYSA-N

Application

Useful in preparation of EDTA conjugates. When conjugated to an affinity probe, e.g., an intercalating agent, chelate can produce interesting site-directed reactions.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M R Ermácora et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(14), 6383-6387 (1992-07-15)
We report the synthesis and evaluation of (EDTA-2-aminoethyl) 2-pyridyl disulfide. By using this easily prepared cysteine-specific hydrophilic reagent, an ethylenediaminetriacetic acid-Fe3+ complex (EDTA-Fe) was covalently attached to a single genetically engineered cysteine residue in staphylococcal nuclease. Upon addition of the
Schultz, P.G., et al.
Journal of the American Chemical Society, 104, 6861-6861 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service