Assay
≥98%
form
powder
solubility
methanol: 10 mg/mL, clear, colorless to light yellow
storage temp.
−20°C
SMILES string
CCCCCCCCC#CCC#CCC#CCCCC(O)=O
InChI
1S/C20H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22/h2-8,11,14,17-19H2,1H3,(H,21,22)
InChI key
OWYNLPMPYBYKJP-UHFFFAOYSA-N
Biochem/physiol Actions
The acetylenic analog of 5,8,11-eicosatrienoic acid is an inhibitor of 12-lipoxygenase and, in MDCK cells, triggers release of Ca2+ from endoplasmic reticulum and other intracellular stores.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
British journal of pharmacology, 134(1), 30-37 (2001-08-28)
1. In the absence of indomethacin, anandamide did not contract the guinea-pig bronchus at concentrations up to 100 microM. In the presence of indomethacin (10 microM), anandamide induced concentration-related contractions with a pEC(50) value of 5.18+/-0.11. It was significantly less
American journal of respiratory and critical care medicine, 202(4), 558-567 (2020-04-24)
Rationale: The role of inspiratory effort still has to be determined as a potential predictor of noninvasive mechanical ventilation (NIV) failure in acute hypoxic de novo respiratory failure.Objectives: To explore the hypothesis that inspiratory effort might be a major determinant
Prostaglandins, 26(6), 1011-1020 (1983-12-01)
The possible involvement of products of the lipoxygenase pathway of arachidonic acid cascade in ovulation was tested by intrabursal injection of nordihydroguaiaretic acid (NDGA); 5, 8, 11-eicosatriynoic acid (5, 8, 11-ETYA), 3 amino-1-(3 trifluromethyphenyl)-2-pyrazoline hydrochloride (BW755c) and (FPL 55712). All
Atherosclerosis, 43(2-3), 143-150 (1982-06-01)
The migration of rat aortic smooth muscle cells was measured in modified Boyden chambers. Smooth muscle cells were motile in vitro and their migration was stimulated (time- and dose-dependently) by a platelet-derived factor. Treatment of platelets with indomethacin resulted in
International journal of immunopharmacology, 10(7), 879-887 (1988-01-01)
The immunomodulatory agent RU 41740 (Biostim), which is derived from Klebsiella pneumoniae, may augment mitogenic responses of purified human blood lymphocytes. In non-purified preparations, however, responses may be sharply reduced due to the fact that Biostim induces monocytes to secrete
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service