DWP96500PLW4-EP
Eppendorf® Deepwell Plate 96/500μL
Protein LoBind, PCR Clean, volume 500 μL, white border with clear wells, pkg of 40 plates (5 bags x 8 plates)
Synonym(s):
cell culture plate, multi well plates, multiple well plates
About This Item
low binding surface
Protein LoBind
white border with clear wells
Recommended Products
sterility
non-sterile
feature
PCR Clean
Protein LoBind
white border with clear wells
packaging
pkg of 40 plates (5 bags x 8 plates)
manufacturer/tradename
Eppendorf® 951032107
availability
available only in North America
technique(s)
PCR: suitable
volume
500 μL
wells
96
binding type
Protein low binding
low binding surface
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- Minimal remaining volume and high uniformity from well to well
- Raised well rims and smooth surface for reliable closing, even with heat sealing
- Easy and reliable stacking
- g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
- High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
- Autoclavable (121?°C, 20?min)
- Available with barcode. Additional details available at: www.eppendorf.com/barcode
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service