Skip to Content
Merck
All Photos(1)

Key Documents

PEA-10

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse unknown/unspecified

packaging

tube of 5 μg 97040310-DNA-5UG
pkg of vial of cells 97040310-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse embryo fibroblast, LRP-deficient, heterozygous

Cell Line Description

The cell line PEA-10 is heterozygous for a disruption of the gene for low density lipoprotein receptor-related protein (LRP). PEA-10 was derived from fibroblasts from day 15 C57BL/6x129 F1 hybrid mouse embryos. Together with the cell lines PEA-13 (Sigma Catalogue number. 97040311) and MEF-1 (Sigma Catalogue number. 98061101) an experimental system is available for the analysis of cellular uptake of functionally diverse ligands and the effect of LRP deficiency. This is of importance since LRP deficient mouse embryos die early during gestation.

Application

Study of LRP and LRP deficiency.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split confleunt cultures 1:6 to 1:10 i.e. seeding 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service