Skip to Content
Merck
All Photos(1)

Key Documents

SVCT

94122105, human breast, Epithelial

Synonym(s):

SCTV, SCVT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

SVCT, 94122105, from human breast

biological source

human breast

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human breast epithelium, SV40 transformed

Cell Line Description

SVCT has been established from human breast epithelial cells being transfected with SV40. The cell line shows an abnormal karyotype and was not tumourigenic in athymic male nude mice. SVCT have been found positive for the expression of keratin 7, 8 and 18 but negative for keratin 19. Epithelial cells, dome formation when confluent.

DNA Profile

Not specified

Culture Medium

DMEM + 2 mM Glutamine + 5μg/ml Hydrocortisone + 10μg/ml Insulin + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4, i.e., seeding at 2-4x10,000 cells/cm2 using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: All customers are required to complete a Cell line Release Authorisation form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service