Skip to Content
Merck
All Photos(1)

Documents

Psi-CRIP

94090901, mouse embryo, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Psi-CRIP, 94090901

biological source

mouse embryo

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Amphotrophic helper-free recombinant retroviral packaging cell line.

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse NIH 3T3 transformed with Moloney murine leukaemia mutants

Cell Line Description

From a set of packaging cell lines useful for generating helper-free recombinant retroviruses with amphotropic host ranges. Two mutant Moloney murine leukaemia virus-derived proviral genomes carrying complementary mutations in the gag-pol or env regions were sequentially introduced into NIH 3T3 cells by co-transformation. Each genome contained a deletion of the Psi sequence necessary for the efficient encapsulation of retroviral genomes into virus particles and additional alterations at the 3′end of the provirus. Psi-CRIP can be used to isolate clones that stably produce high titres (1000,000cfu/ml) of recombinant retroviruses with amphotropic host ranges. Viral producers derived from Psi-CRIP do not transfer the packaging function or yield helper virus.

Application

Amphotropic helper-free recombinant retroviral packaging

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (NB. The results obtained by the depositor were gained using 10% Calf Serum (CS)).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5-10% CO2, 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service