Skip to Content
Merck
All Photos(1)

Documents

LL47 (MaDo)

90102538, human lung, Fibroblast-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LL47 (MaDo), 90102538

biological source

human lung

growth mode

Adherent

karyotype

2n = 46; diploid

morphology

Fibroblast-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human diploid lung

Cell Line Description

Derived from normal lung tissue obtained from a 16 year old male with osteogenic sarcoma. Ethnicity: Black.

Application

Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 7,12
D13S317: 9,11
D16S539: 11
D5S818: 12,13
D7S820: 8,10
THO1: 7,9.3
TPOX: 9,10
vWA: 14,16

Culture Medium

Coon′s modified Ham′s F12 + Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. A further 15 population doublings expected.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service