Skip to Content
Merck
All Photos(1)

Documents

YTS 121.5.2

87072283

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

technique(s)

cell culture | mammalian: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−196°C

Other Notes

Cultures from HPA Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service