Skip to Content
Merck
All Photos(1)

Key Documents

3T6 Swiss Albino

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 86120801, mouse embryo, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

mouse embryo

packaging

tube of 5 μg 86120801-DNA-5UG
pkg of vial of cells 86120801-1VL

growth mode

Adherent

karyotype

2n = 40

morphology

Fibroblast

products

Collagen and hyaluronic acid

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

General description

Product has lost contact inhibition

Cell Line Origin

Mouse Swiss Albino embryo fibroblast

Cell Line Description

Derived from Swiss mouse embryos this cell line secretes collagen and hyaluronic acid. Unlike 3T3, the 3T6 line is not contact inhibited.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:20 i.e. seeding at 5x1,000 to 3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service