Skip to Content
Merck
All Photos(1)

Key Documents

UM-UC-13

8090508, human bladder, Epithelial

Synonym(s):

Human UM-UC-13, UM-UC-13 Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

UM-UC-13, 08090508

biological source

human bladder

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

metastasis

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human transitional cell carcinoma lymphatic metastasis

Cell Line Description

UM-UC-13 was produced from a lymphatic metastasis from a transitional cell carcinoma of the bladder. UM-UC-13 produced tumours in female athymic nude mice after 36 days. Susceptibility to adenoviral mediated gene transduction and expression of p53, p16, retinoblastoma protein and coxsackie adenovirus receptor were studied.

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 10,12
D13S317: 8,12
D16S539: 11
D5S818: 9,13
D7S820: 8,9
THO1: 9
TPOX: 8
vWA: 16,18

Culture Medium

EMEM (EBSS) + 2 mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37 °C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service