Skip to Content
Merck
All Photos(1)

Key Documents

CHP-100

6122001

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207

Product Name

CHP-100, 06122001

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see CHP-100

Cell Line Origin

Human neuroblastoma epidural tumour of spine

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service