Skip to Content
Merck
All Photos(1)

Key Documents

C9131

Sigma-Aldrich

Sodium N-cyclohexylsulfamate

Synonym(s):

N-Cyclohexylsulfamic acid sodium salt, Cyclamic acid sodium salt, Cyclohexanesulfamic acid sodium salt, Sodium cyclamate, Sodium cyclohexanesulfamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NHSO3Na
CAS Number:
Molecular Weight:
201.22
Beilstein:
4166868
EC Number:
MDL number:
UNSPSC Code:
12352103

mp

>300 °C (lit.)

SMILES string

[Na+].[O-]S(=O)(=O)NC1CCCCC1

InChI

1S/C6H13NO3S.Na/c8-11(9,10)7-6-4-2-1-3-5-6;/h6-7H,1-5H2,(H,8,9,10);/q;+1/p-1

InChI key

UDIPTWFVPPPURJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Phenylsulfamate sweet tastant detectable by humans. ′First generation′ sweetener studied in animal models for its carcinogenic potential.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Salas et al.
Analytical and bioanalytical chemistry, 407(15), 4277-4285 (2014-11-28)
Artificial sweeteners are food additives employed as sugar substitutes which are now considered to be emerging organic contaminants. In the present study, a method is developed for the determination of a group of artificial sweeteners in environmental waters. Considering the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service