Skip to Content
Merck
All Photos(2)

Key Documents

C7438

Sigma-Aldrich

Chaperonin 10 from Escherichia coli

≥95.0% (SDS-PAGE), recombinant, expressed in E. coli overproducing strain, lyophilized powder

Synonym(s):

GroES

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
€843.00

€843.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
€843.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

€843.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Escherichia coli

Quality Level

recombinant

expressed in E. coli overproducing strain

Assay

≥95.0% (SDS-PAGE)

form

lyophilized powder

technique(s)

activity assay: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... HSPE1(3336)

Biochem/physiol Actions

Chaperonin60 (GroEL) and chaperonin10 (GroES) belong to the ubiquitous family of heat-shock molecular chaperones found in prokaryotes and in eukaryotic organelles. The chaperonins assist the folding of nascent, organelle-imported or stress-destabilized polypeptides. In vitro, purified GroEL together with purified GroES in the presence of Mg-ATP facilitate refolding and reactivation of denatured proteins, e.g., the photosynthetic enzyme rubisco and the mitochondrial enzyme rhodanese.
The folding activity of a 1:1 molar mixture of GroEL and GroES was tested using urea-denatured rhodanese. At least 2-fold reactivation of rhodanese over the spontaneous reactivation was obtained.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing Tris buffer salts, potassium chloride, dithiothreitol and trehalose as stabilizer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service