Skip to Content
Merck
All Photos(1)

Key Documents

C7339

Sigma-Aldrich

Monoclonal Anti-Cyclin D2 antibody produced in mouse

clone DCS-3, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

clone

DCS-3, monoclonal

mol wt

antigen 33-35 kDa (and several lower and higher MW proteins)

contains

15 mM sodium azide

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1:400 using a cultured human tumor cell line extract

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CCND2(894)
mouse ... Ccnd2(12444)

Specificity

The antibody is useful in antibody-mediated knock-out experiments. The product does not cross-react with other D-type cyclins.

Immunogen

recombinant human cyclin D2 protein.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathalie M Fiaschi-Taesch et al.
Diabetes, 59(8), 1926-1936 (2010-07-30)
Most knowledge on human beta-cell cycle control derives from immunoblots of whole human islets, mixtures of beta-cells and non-beta-cells. We explored the presence, subcellular localization, and function of five early G1/S phase molecules-cyclins D1-3 and cdk 4 and 6-in the
J Lukas et al.
Oncogene, 10(11), 2125-2134 (1995-06-01)
To explore regulation and function of cyclin D2, a candidate cell cycle-regulatory proto-oncogene, we examined subcellular localisation, cell type- and cell cycle-dependent expression, and requirement of cyclin D2 protein for G1 progression, in a panel of 40 human normal and
Ningli Cheng et al.
PloS one, 3(4), e1911-e1911 (2008-04-03)
The effect of Bcl-2 on oncogenesis is complex and expression may either delay or accelerate oncogenesis. The pro-oncogenic activity is attributed to its well characterized anti-apoptotic function while the anti-oncogenic function has been attributed to its inhibition of cellular proliferation.
Kikumi Hata et al.
Journal of immunology (Baltimore, Md. : 1950), 173(4), 2453-2461 (2004-08-06)
The engagement of membrane-bound Igs (mIgs) results in growth arrest, accompanied by apoptosis, in the WEHI-231 murine B lymphoma cells, a cell line model representative of primary immature B cells. Inhibitor of differentiation (Id) proteins, members of the helix-loop-helix protein
Amber Yasmeen et al.
Journal of oncology, 2010 (2010-09-24)
In this study, we compared the effects of SKI-606 with Iressa, Src/Abl and EGF-R kinase inhibitors, respectively, on selected parameters in HeLa and SiHa cervical cancer cell lines, which express E6/E7 oncoproteins of high-risk HPV types 18 and 16, respectively.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service