Skip to Content
Merck
All Photos(1)

Key Documents

C6922

Sigma-Aldrich

Cyclohexylacetyl-Phe-Arg-Ser-Val-Gln amide

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H58N10O8
CAS Number:
Molecular Weight:
758.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C(NC(=O)C(CO)NC(=O)C(CCCNC(N)=N)NC(=O)C(Cc1ccccc1)NC(=O)CC2CCCCC2)C(=O)NC(CCC(N)=O)C(N)=O

InChI

1S/C36H58N10O8/c1-21(2)30(35(54)43-24(31(38)50)15-16-28(37)48)46-34(53)27(20-47)45-32(51)25(14-9-17-41-36(39)40)44-33(52)26(18-22-10-5-3-6-11-22)42-29(49)19-23-12-7-4-8-13-23/h3,5-6,10-11,21,23-27,30,47H,4,7-9,12-20H2,1-2H3,(H2,37,48)(H2,38,50)(H,42,49)(H,43,54)(H,44,52)(H,45,51)(H,46,53)(H4,39,40,41)

InChI key

CQRVUZFZOUAQCT-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Okunishi et al.
Advances in experimental medicine and biology, 247B, 23-28 (1989-01-01)
Kininogen sequence analogs containing amino acid residues around the Arg-Ser cleavage site of bovine kininogens were prepared with bulky aliphatic residues in P3 position. KKI-7 (containing a cyclohexylacetyl group) and KKI-8 (containing an adamantaneacetyl group) both inhibited human urinary kallikrein
The design of specific inhibitors of tissue kallikrein and their effect on the blood pressure of the rat.
J Burton et al.
Advances in experimental medicine and biology, 247B, 9-13 (1989-01-01)
P Rebuffat et al.
Histology and histopathology, 15(2), 441-444 (2000-05-16)
The effects on the pituitary-adrenocortical functions of the prolonged (7-day) blockade of endogenous bradykinin (BK) synthesis, obtained by the administration of the kallikrein inhibitor (K-I) cyclohexylacetyl-Phe-Arg-Ser-Val-Gln amide, were investigated in the rat. K-I treatment did not cause significant changes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service