Skip to Content
Merck
All Photos(1)

Key Documents

C2747

Sigma-Aldrich

Anti-CPSF3 (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CPSF73, Anti-Cleavage and Polyadenylation Specificity Factor 3, Anti-YSH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

human

technique(s)

western blot: 1:250-1:500 using K-562 lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPSF3(51692)

General description

Caspase-3 is a caspase protein that interacts with caspase-8 and caspase-9. It is encoded by the CASP3 gene. This gene is a member of the metallo-β-lactamase family and is referred as CPSF73 and CPSF-73. CPSF73 is a 73kDa subunit and is the pre-mRNA 3′-end-processing endonuclease.

Application

Anti-CPSF3 (C-terminal) antibody produced in rabbit is suitable for western blotting at a dilution of 1:250-1:500 using K-562 lysate.

Biochem/physiol Actions

CPSF3 is induced by interaction between CSR1 and CPSF3 and is translocated from the nucleus to the cytoplasm, resulting in inhibition of polyadenylation. Down-regulation of CPSF3 using small interfering RNA induces cell death. CPSF73 mediates cleavage coupled to polyadenylation and histone pre-mRNA processing. HIV-1 Tat protein interacts with CPSF-73 and counteracts its repressive activity on the HIV-1 LTR promoter. During the cleavage and polyadenylation, CPSF plays a central role in processing the reaction.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laureano de la Vega et al.
Journal of molecular biology, 372(2), 317-330 (2007-08-03)
Gene expression in eukaryotes requires the post-transcriptional cleavage of mRNA precursors into mature mRNAs. The cleavage and polyadenylation specificity factor (CPSF) is critical for this process and its 73 kDa subunit (CPSF-73) mediates cleavage coupled to polyadenylation and histone pre-mRNA
Corey R Mandel et al.
Nature, 444(7121), 953-956 (2006-11-28)
Most eukaryotic messenger RNA precursors (pre-mRNAs) undergo extensive maturational processing, including cleavage and polyadenylation at the 3'-end. Despite the characterization of many proteins that are required for the cleavage reaction, the identity of the endonuclease is not known. Recent analyses
A Jenny et al.
Science (New York, N.Y.), 274(5292), 1514-1517 (1996-11-29)
The 3' ends of most eukaryotic messenger RNAs are generated by endonucleolytic cleavage and polyadenylation. In mammals, the cleavage and polyadenylation specificity factor (CPSF) plays a central role in both steps of the processing reaction. Here, the cloning of the
Z-H Zhu et al.
Oncogene, 28(1), 41-51 (2008-09-23)
CSR1 (cellular stress response 1), a newly characterized tumor-suppressor gene, undergoes hypermethylation in over 30% of prostate cancers. Re-expression of CSR1 inhibits cell growth and induces cell death, but the mechanism by which CSR1 suppresses tumor growth is not clear.
Lee Davidson et al.
Genes & development, 28(4), 342-356 (2014-01-31)
3' end formation of pre-mRNAs is coupled to their transcription via the C-terminal domain (CTD) of RNA polymerase II (Pol II). Nearly all protein-coding transcripts are matured by cleavage and polyadenylation (CPA), which is frequently misregulated in disease. Understanding how

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service