Skip to Content
Merck
All Photos(1)

Key Documents

C009

Sigma-Aldrich

McN-A-343

Synonym(s):

(4-Hydroxy-2-butynyl)-1-trimethyl­ammonium-3-chloro­carbanilate chloride, 4-[N-(3-Chlorophenyl)carbamoyloxy]-2-­butynyl­trimethyl­ammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cl2N2O2
CAS Number:
Molecular Weight:
317.21
MDL number:
UNSPSC Code:
12352200

color

white

SMILES string

[Cl-].C[N+](C)(C)CC#CCOC(=O)Nc1cccc(Cl)c1

InChI

1S/C14H17ClN2O2.ClH/c1-17(2,3)9-4-5-10-19-14(18)16-13-8-6-7-12(15)11-13;/h6-8,11H,9-10H2,1-3H3;1H

InChI key

CXFZFEJJLNLOTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

M1 muscarinic acetylcholine receptor agonist.

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service