Skip to Content
Merck
All Photos(1)

Key Documents

BMOPTIONC-PREC

B Medical Temperature Recorder 7 Day, 5", Abacus For LF260, L290, L380, P260, P290, P380 (+4C)

Accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025

packaging

pack of 1 ea

manufacturer/tradename

B Medical Systems

shipped in

ambient

storage temp.

room temp

General description

Temperature Recorder 7 Day, 5", Battery Operated - Abacus For LF260, L290, L380, P260, P290, P380 (+4C)

Application

Accessory

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service