Skip to Content
Merck
All Photos(1)

Key Documents

B9305

Sigma-Aldrich

BW 245C

≥98% (HPLC), solid

Synonym(s):

3-(3-Cyclohexyl-3-hydroxypropyl)-2,5-dioxo-(R*,S*)-(±)-4-imidazolineheptanioc acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H32N2O5
CAS Number:
Molecular Weight:
368.47
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble 10 mg/mL

originator

GlaxoSmithKline

storage temp.

−20°C

SMILES string

O[C@H](CCN1[C@@H](CCCCCCC(O)=O)C(=O)NC1=O)C2CCCCC2

InChI

1S/C19H32N2O5/c22-16(14-8-4-3-5-9-14)12-13-21-15(18(25)20-19(21)26)10-6-1-2-7-11-17(23)24/h14-16,22H,1-13H2,(H,23,24)(H,20,25,26)/t15-,16+/m0/s1

InChI key

ZIDQIOZJEJFMOH-JKSUJKDBSA-N

Gene Information

human ... PTGDR(5729)

Biochem/physiol Actions

BW 245C was investigated for the affinities potencies, and intrinsic activities in comparison with other natural and synthetic prostanoids using endogenous receptors. The rank order of compound affinities at the DP receptor was SQ27986 (K(i) = 10 +/- 2 nM) > RS93520 = ZK110841 = BW245C (K(i) = 23-26 nM) > ZK118182 (K(i) = 50 +/- 9 nM) > PGD(2) (K(i) = 80 +/- 5 nM). DP receptor agonists produced cAMP in embryonic bovine tracheal fibroblasts with different potencies (EC(50) values in nM): ZK118182 (18 +/- 6), RS93520 (28 +/- 6), SQ27986 (29 +/- 7), ZK110841 (31 +/- 7), BW245C (53 +/- 16), and PGD(2) (98 +/- 10). BW245C was more efficacious and RS93520 was less efficacious.

Features and Benefits

This compound is featured on the Prostanoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xibin Liang et al.
Journal of neurochemistry, 92(3), 477-486 (2005-01-22)
Cyclo-oxygenases (COXs) catalyze the first committed step in the synthesis of the prostaglandins PGE(2), PGD(2), PGF(2alpha), PGI(2) and thomboxane A(2). Expression and enzymatic activity of COX-2, the inducible isoform of COX, are observed in several neurological diseases and result in
N A Sharif et al.
The Journal of pharmacology and experimental therapeutics, 293(2), 321-328 (2000-04-25)
The prostanoid receptor-subtype binding affinities, selectivities, potencies, and intrinsic activities of four natural prostanoids and six synthetic DP class prostanoids were determined using binding and functional assays with endogenous receptors. SQ27986 exhibited the highest affinity for the human platelet DP
Alicja Jozkowicz et al.
Antioxidants & redox signaling, 10(12), 2035-2046 (2008-07-31)
15-Deoxy-Delta(12,14)-prostaglandin-J(2) (15d-PGJ(2)) is a cyclopentenone prostaglandin regarded as antiinflammatory mediator, which can act through peroxisome proliferator-activated receptor-gamma (PPARgamma) or through G protein-coupled surface receptors. It has been demonstrated that 15d-PGJ(2) potently increases the generation of interleukin-8 (IL-8) in human microvascular
C Yoshimura-Uchiyama et al.
Clinical and experimental allergy : journal of the British Society for Allergy and Clinical Immunology, 34(8), 1283-1290 (2004-08-10)
Both prostaglandin (PG) D receptor (DP) and CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells)/DP2 are high-affinity receptors for PGD2. Previous studies have demonstrated that PGD2 enhances releasability and induces CRTH2/DP2-mediated migration in human basophils, but the precise effects of
Sybille van den Brule et al.
The Journal of pharmacology and experimental therapeutics, 335(2), 472-479 (2010-08-20)
Prostaglandin (PG) D(2) exerts contrasting activities in the inflamed lung via two receptors, the D prostanoid receptor (DP) and the chemoattractant receptor-homologous molecule expressed on T helper 2 lymphocytes. DP activation is known mainly to inhibit proinflammatory cell functions. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service