B5256
DL-α-O-Benzylglycerol
≥98%
Synonym(s):
1-O-Benzyl-rac-glycerol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥98%
storage temp.
2-8°C
SMILES string
OCC(O)COCc1ccccc1
InChI
1S/C10H14O3/c11-6-10(12)8-13-7-9-4-2-1-3-5-9/h1-5,10-12H,6-8H2
InChI key
LWCIBYRXSHRIAP-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Aliphatic diether analogs of glyceride-derived lipids. I. Synthesis of d-alpha,beta-dialkyl glyceryl ethers.
Biochemistry, 2, 394-397 (1963-03-01)
Bioorganic & medicinal chemistry, 17(1), 411-417 (2008-01-10)
The in vitro inhibitory effects and in silico docking energies of 18 compounds with respect to aquaporin 4 (AQP4) were investigated. More than half of the compounds tested showed inhibitory activity in the in vitro functional assay and included the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service