Skip to Content
Merck
All Photos(1)

Key Documents

B3183

Sigma-Aldrich

Anti-BID antibody produced in rabbit

Cleavage site [59/60] specific, affinity isolated antibody, buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 15 kDa

species reactivity

mouse

technique(s)

microarray: suitable
western blot: suitable using mouse L929 cells treated with TNF-α

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Bid(12122)

Immunogen

synthetic peptide corresponding to N-terminal of cleavage site [59/60] of mouse BID.

Application

Polyclonal anti-BID antibody produced in rabbit is suitable for western blotting by using mouse L929 cells treated with TNF-α. It is also suitable for microarray.

Biochem/physiol Actions

The protein encoded by this gene is a member of the BCL-2 family of cell death regulators and is localized on to chromosome 22q11.2. BID is activated by caspase 8 in response to Fas/TNF-R1 receptor activation. Activated BID then translocated to mitochondria where it induces cytochrome c release, which in turn activates downstream caspases. It is a BH3 interacting death domain that heterodimerizes with either agonist BAX or antagonist BCL2. BID initiates apoptosis by binding to regulatory sites on prosurvival BCL2 proteins to directly neutralize their function. Additionally, BID interacts with cathepsins play an important role in the actions of Camptothecin on breast cancer cells.

Physical form

Supplied as a solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1.0 mg/ml BSA (IgG and protease free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T K Footz et al.
Genomics, 51(3), 472-475 (1998-08-29)
Cat eye syndrome (CES) is associated with a duplication of a segment of human chromosome 22q11.2. Only one gene, ATP6E, has been previously mapped to this duplicated region. We now report the mapping of the human homologue of the apoptotic
M Lamparska-Przybysz et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 56 Suppl 3, 159-179 (2005-08-04)
The details of molecular switching points between apoptosis and autophagy in tumor cells have still not been fully elucidated. This study focused on the role of cathepsin B and its substrate, BID as molecular links between apoptosis and autophagy in
H Li et al.
Cell, 94(4), 491-501 (1998-09-04)
We report here that BID, a BH3 domain-containing proapoptotic Bcl2 family member, is a specific proximal substrate of Casp8 in the Fas apoptotic signaling pathway. While full-length BID is localized in cytosol, truncated BID (tBID) translocates to mitochondria and thus
Vijay Agarwal et al.
International journal of oncology, 42(3), 1088-1092 (2013-01-23)
We have previously shown that specific COX-2 inhibitors, including DuP 697, have anti-proliferative effects on mesothelioma cells and potentiate the cytotoxicity of pemetrexed. Here, we used a novel proteomic approach to explore the mechanism of action of this agent. COX-2-positive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service