B2766
Anti-Mouse IgA (α-chain specific)−Biotin antibody produced in goat
affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
Recommended Products
biological source
goat
conjugate
biotin conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
direct ELISA: 1:20,000
storage temp.
2-8°C
Physical form
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Noah W Palm et al.
Cell, 158(5), 1000-1010 (2014-08-30)
Specific members of the intestinal microbiota dramatically affect inflammatory bowel disease (IBD) in mice. In humans, however, identifying bacteria that preferentially affect disease susceptibility and severity remains a major challenge. Here, we used flow-cytometry-based bacterial cell sorting and 16S sequencing
Sule Karaman et al.
Molecular biotechnology, 32(1), 31-42 (2005-12-31)
Enterotoxigenic strains of Escherichia coli produce a heat-labile holotoxin (LT), which causes diarrhea. We engineered corn seeds to produce LT-B, the nontoxic subunit of LT, to serve as a plant-derived vaccine to traveler's diarrhea and as an adjuvant for co-administered
Marcel Maier et al.
Vaccine, 23(44), 5149-5159 (2005-08-02)
Abeta vaccination or passive transfer of human-specific anti-Abeta antibodies are approaches under investigation to prevent and/or treat Alzheimer's disease (AD). Successful active Abeta vaccination requires a strong and safe adjuvant to induce anti-Abeta antibody formation. We compared the adjuvants monophosphoryl
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service