Skip to Content
Merck
All Photos(1)

Key Documents

B0561

Sigma-Aldrich

Monoclonal Anti-BUB1 antibody produced in mouse

2 mg/mL, clone 14H5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Budding Uninhibited By Benzimidazoles 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

14H5, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

2 mg/mL

technique(s)

immunocytochemistry: 4-8 μg/mL using HeLa cells treated with 20 μg/mL nacodazol
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BUB1(699)

General description

BUB1 is spindle checkpoint kinase that modulates kinetochore-independent segregation of chromosomes and cell proliferation. This kinase regulates the phosphorylation of MAD1 and itself. As a result, BUB1 mediates chromosome-spindle attachments during mitosis. BUB1 also phosphorylates Cdc20 which subsequently facilitates accurate checkpoint signaling . Monoclonal Anti-BUB1 antibody is specific for human BUB1 (approx. 150 kDa).

Immunogen

recombinant human BUB1.

Application

Monoclonal Anti-BUB1 antibody is suitable for use in western blot, indirect ELISA, microarray, immunocytochemistry (4-8 μg/mL using HeLa cells treated with 20 μg/mL nacodazol) and immunoprecipitation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhanyun Tang et al.
Molecular cell, 16(3), 387-397 (2004-11-05)
To ensure the fidelity of chromosome segregation, the spindle checkpoint blocks the ubiquitin ligase activity of APC/C(Cdc20) in response to a single chromatid not properly attached to the mitotic spindle. Here we show that HeLa cells depleted for Bub1 by
B Ouyang et al.
Cell growth & differentiation : the molecular biology journal of the American Association for Cancer Research, 9(10), 877-885 (1998-10-28)
Eukaryotic cells have evolved a mechanism that delays the onset of anaphase until chromosomes are properly positioned on the spindle. To understand the molecular basis of such surveillance mechanism in human cells, we have cloned a full-length cDNA encoding a
T W Seeley et al.
Biochemical and biophysical research communications, 257(2), 589-595 (1999-04-13)
The BUB/MAD signaling pathway monitors attachment of chromosomes to spindle poles in mitotic cells. Mutations of the human BUB1 locus were identified in cancer cells exhibiting an unstable chromosomal complement. We report that the human BUB3 gene maps to a
Christiane Klebig et al.
The Journal of cell biology, 185(5), 841-858 (2009-06-03)
The kinetochore-bound protein kinase Bub1 performs two crucial functions during mitosis: it is essential for spindle checkpoint signaling and for correct chromosome alignment. Interestingly, Bub1 mutations are found in cancer tissues and cancer cell lines. Using an isogenic RNA interference
Min Wu et al.
Nature communications, 10(1), 273-273 (2019-01-19)
Faithful chromosome segregation requires proper chromosome congression at prometaphase and dynamic maintenance of the aligned chromosomes at metaphase. Chromosome missegregation can result in aneuploidy, birth defects and cancer. The kinetochore-bound KMN network and the kinesin motor CENP-E are critical for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service