Skip to Content
Merck
All Photos(1)

Documents

B0431

Sigma-Aldrich

Anti-Bcl-10 (MK-17) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~32 kDa

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:8,000 using trypsin-treated sections of rat spleen
immunoprecipitation (IP): 2.5-5 μg using lysate from 4-6x105 human THP-1/Raji cells
microarray: suitable
western blot: 1:500 using whole extract of human THP-1/ Raji cells and mouse NIH3T3 fibroblasts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... BCL10(8915)
mouse ... Bcl10(12042)
rat ... Bcl10(83477)

Immunogen

synthetic peptide corresponding to the human Bcl-10 sequence (amino acids 1-17) conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service