Skip to Content
Merck
All Photos(1)

Key Documents

B010

Sigma-Aldrich

Benextramine tetrahydrochloride

Synonym(s):

BHC, Dithiobis(N-[N-(2-methoxybenzyl)-6-aminohexyl]-2-aminoethane), Dithiobis(N-[N-(2-methoxybenzyl)-6-aminohexyl]-2-aminoethane);, N,N′′-(Dithiodi-2,1-ethanediyl)bis[N′-(2-methoxyphenylmethyl)-1,6-hexanediamine] dihydrochloride, N,N′-Bis(6-[o-Methoxybenzylamino]hexyl)cystamine, N,N′-Bis(6-[o-methoxybenzylamino]hexyl)cystamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H54N4O2S2 · 4HCl
CAS Number:
Molecular Weight:
736.77
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

Cl.COc1ccccc1CNCCCCCCNCCSSCCNCCCCCCNCc2ccccc2OC

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service