Key Documents
AXYTE1004BLRS
MAXYMum Recovery® pipette tips for Eppendorf® and Socorex pipettors
capacity 1000 μL, blue, sterile
Synonym(s):
pipet tips
Select a Size
1 PKG
€430.00
€430.00
Please contact Customer Service for Availability
Select a Size
Change View
1 PKG
€430.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
1000 μL
sterility:
γ-irradiated
sterile
feature:
autoclavable
packaging:
case of 3456 tips racked
manufacturer/tradename:
Axygen Inc
Corning
€430.00
Please contact Customer Service for Availability
material
blue
sterility
γ-irradiated
sterile
feature
autoclavable
packaging
case of 3456 tips racked
manufacturer/tradename
Axygen Inc
Corning
capacity
1000 μL
volume
1000 μL
Related Categories
Other Notes
Browse all pipette tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)