Skip to Content
Merck
All Photos(1)

Key Documents

AXYPCR96LC480CNF

Axygen 96 well PCR plate for Roche 480 LightCycler

clear plate

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
material:
clear
size:
96 wells
Product AXYPCR96LC480CNF is not currently sold in your country. Contact Technical Service

material

clear

manufacturer/tradename

Corning PCR96LC480CNF

size

96 wells

color

clear plate

suitability

suitable for PCR


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service